Thermo Scientific Chemicals

2-Amino-5-nitrobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A10426.18
50 g, Each
Thermo Scientific Chemicals

2-Amino-5-nitrobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A10426.18
50 g, Each
Quantity
Catalog number: A10426.18
also known as A10426-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
616-79-5
Form
Powder or lumps
Appearance (Color)
Yellow to orange
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

2-Amino-5-nitrobenzoic acid, is used as an intermediate for dyes and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-nitrobenzoic acid, is used as an intermediate for dyes and pharmaceuticals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatabile to strong oxidizing agents. Stable under ordinary conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text