Thermo Scientific Chemicals

Aminoacetaldehyde diethyl acetal, 98%, Thermo Scientific Chemicals

Catalog number: A10427.22
100 g, Each
Thermo Scientific Chemicals

Aminoacetaldehyde diethyl acetal, 98%, Thermo Scientific Chemicals

Catalog number: A10427.22
100 g, Each
Quantity
Catalog number: A10427.22
also known as A10427-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
645-36-3
IUPAC Name
2,2-diethoxyethan-1-amine
Molecular Formula
C6H15NO2
InChI Key
HJKLEAOXCZIMPI-UHFFFAOYSA-N
SMILES
CCOC(CN)OCC
Form
Liquid
Comment
Material sourced in the U.S. and in other countries
Refractive Index
1.4155-1.4195 @ 20°C (non-U.S. sourced material)
Appearance (Color)
Clear colorless to pale yellow
Infrared spectrum
Conforms (U.S. sourced material)

Description

Aminoacetaldehyde diethyl acetal, is used as an agrochemical intermediate, pharmaceutical, syntheses material intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aminoacetaldehyde diethyl acetal, is used as an agrochemical intermediate, pharmaceutical, syntheses material intermediate.

Solubility
Soluble in water.

Notes
Moisture Sensitive. Store in a cool, dry place. Store in a tightly closed container.Take measures to prevent the build up of electrostatic charge. Stable under recommended storage conditions. Incompatabile to strong oxidizing agents, strong acids, acid chlorides, acid anhydrides, Carbon dioxide (CO2).
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text