Thermo Scientific Chemicals

Caffeine, 99%, Thermo Scientific Chemicals

Catalog number: A10431.22
100 g, Each
Thermo Scientific Chemicals

Caffeine, 99%, Thermo Scientific Chemicals

Catalog number: A10431.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A10431.22
also known as A10431-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
58-08-2
IUPAC Name
1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Molecular Formula
C8H10N4O2
InChI Key
RYYVLZVUVIJVGH-UHFFFAOYSA-N
SMILES
CN1C=NC2=C1C(=O)N(C)C(=O)N2C
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Appearance (Color)
White to pale cream
Identification (FTIR)
Conforms

Description

This material is traded for research purposes ONLY. Thermo Fisher does NOT certify this material as suitable for pharmaceutical, cosmetic, veterinary or food use. Caffeine has inhibitory action against lung and colon tumorigenses and virus induction.Caffeine is used to treat and prevent bronchopulmonary dysplasia in premature infants. It is also used for the treatment of apnea of prematurity and orthostatic hypotension. It is widely used as an additive in food, cosmetics, pharmaceuticals and beverages. It confers a protective effect against some diseases including Parkinson's disease and certain types of cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in pyrrole, tetrahydrofuran+ water mixture and ethyl acetate. Slightly soluble in water and ethanol.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text