4'-Hydroxy-3'-methoxyacetophenone, 98%, Thermo Scientific Chemicals
4'-Hydroxy-3'-methoxyacetophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4'-Hydroxy-3'-methoxyacetophenone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
500 g
Catalog number A10439.18
also known as A10439-18
Price (USD)/ Each
41.70
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
41.70
Add to cart
4'-Hydroxy-3'-methoxyacetophenone, 98%, Thermo Scientific Chemicals
Catalog numberA10439.18
Price (USD)/ Each
41.70
-
Add to cart
Chemical Identifiers
CAS498-02-2
IUPAC Name1-(4-hydroxy-3-methoxyphenyl)ethan-1-one
Molecular FormulaC9H10O3
InChI KeyDFYRUELUNQRZTB-UHFFFAOYSA-N
SMILESCOC1=CC(=CC=C1O)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)> 97.5%
Formcrystalline powder
Appearance (Color)White to cream or pale yellow to pale brown
4'-Hydroxy-3'-methoxyacetophenone is used as an inhibitor of nicotinamide adenine dinucleotide phosphate (NADPH) oxidase, which is an enzyme responsible for reactive oxygen species production. It is also useful in the treatment of various inflammatory diseases and atherosclerosis. Further, it shows an anti-inflammatory activity and improves endothelial function by reducing oxidative stress. In addition to this, it is used for studying nitric oxide regulated processes in plants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in ethanol, chloroform, ether, hot water, benzene, ethyl acetate and dimethyl sulfoxide. Insoluble in petroleum ether.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Harman-Ware, A. E.; Crocker, M.; Pace, R. B.; Placido, A.; Morton III, S.; DeBolt, S. Characterization of Endocarp Biomass and Extracted Lignin Using Pyrolysis and Spectroscopic Methods. Bioenergy Res. 2015, 8 (1), 350-368.
  2. McKay, D. L.; Chen, C. Y. O.; Zampariello, C. A.; Blumberg, J. B. Flavonoids and phenolic acids from cranberry juice are bioavailable and bioactive in healthy older adults. Food Chem. 2015, 168, 233-240.