Thermo Scientific Chemicals

4'-Hydroxy-3'-methoxyacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: A10439.36
500 g, Each
Thermo Scientific Chemicals

4'-Hydroxy-3'-methoxyacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: A10439.36
500 g, Each
Quantity
Catalog number: A10439.36
also known as A10439-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
498-02-2
IUPAC Name
1-(4-hydroxy-3-methoxyphenyl)ethan-1-one
Molecular Formula
C9H10O3
InChI Key
DFYRUELUNQRZTB-UHFFFAOYSA-N
SMILES
COC1=CC(=CC=C1O)C(C)=O
Form
crystalline powder
Appearance (Color)
White to cream or pale yellow to pale brown
Assay (GC)
> 97.5%

Description

4'-Hydroxy-3'-methoxyacetophenone is used as an inhibitor of nicotinamide adenine dinucleotide phosphate (NADPH) oxidase, which is an enzyme responsible for reactive oxygen species production. It is also useful in the treatment of various inflammatory diseases and atherosclerosis. Further, it shows an anti-inflammatory activity and improves endothelial function by reducing oxidative stress. In addition to this, it is used for studying nitric oxide regulated processes in plants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in ethanol, chloroform, ether, hot water, benzene, ethyl acetate and dimethyl sulfoxide. Insoluble in petroleum ether.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text