Thermo Scientific Chemicals

3,5-Dihydroxytoluene monohydrate, 97%, Thermo Scientific Chemicals

Catalog number: A10440.14
25 g, Each
Thermo Scientific Chemicals

3,5-Dihydroxytoluene monohydrate, 97%, Thermo Scientific Chemicals

Catalog number: A10440.14
25 g, Each
Quantity
Catalog number: A10440.14
also known as A10440-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6153-39-5
IUPAC Name
5-methylbenzene-1,3-diol hydrate
Molecular Formula
C7H10O3
InChI Key
NBKPNAMTHBIMLA-UHFFFAOYSA-N
SMILES
O.CC1=CC(O)=CC(O)=C1
Assay (GC)
≥96.0%
Melting Point
55-65?C
Form
Crystals or crystalline powder
Water Content (Karl Fischer Titration)
10-15%
Appearance (Color)
White to pale brown to pink-brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text