Thermo Scientific Chemicals

Dicyandiamide, 99%, Thermo Scientific Chemicals

Catalog number: A10451.36
500 g, Each
Thermo Scientific Chemicals

Dicyandiamide, 99%, Thermo Scientific Chemicals

Catalog number: A10451.36
500 g, Each
Quantity
Catalog number: A10451.36
also known as A10451-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
461-58-5
IUPAC Name
N''-cyanoguanidine
Molecular Formula
C2H4N4
InChI Key
QGBSISYHAICWAH-UHFFFAOYSA-N
SMILES
NC(N)=NC#N
Form
Crystals or powder or crystalline powder or flakes
Melting Point (clear melt)
206-213?C
Appearance (Color)
White
Identification (FTIR)
Conforms
Assay from Supplier's CofA
≥98.5%

Description

Dicyandiamide is used in the synthesis of barbiturates. It is used as a stabilizer of ammonium dinitramide melt. It is used as hardener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dicyandiamide is used in the synthesis of barbiturates. It is used as a stabilizer of ammonium dinitramide melt. It is used as hardener.

Solubility
Soluble in Water (32g/L ) at 20°C

Notes
Store at 2-8°C. Incompatible with strong acids, strong oxidizing agents, strong acids, strong bases, ammonium nitrate, potassium chlorates.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text