Sesamol, 98%, Thermo Scientific Chemicals
Sesamol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sesamol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A10453.09
also known as A10453-09
Price (USD)/ Each
49.40
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
49.40
Add to cart
Sesamol, 98%, Thermo Scientific Chemicals
Catalog numberA10453.09
Price (USD)/ Each
49.40
-
Add to cart
Chemical Identifiers
CAS533-31-3
IUPAC Name2H-1,3-benzodioxol-5-ol
Molecular FormulaC7H6O3
InChI KeyLUSZGTFNYDARNI-UHFFFAOYSA-N
SMILESOC1=CC=C2OCOC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)61-66?C
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)White to brown
Assay (GC)≥97.5%
View more
Sesamol has good antioxidant activity. Sesamol has potential protective effects against free radicals as well as some antifungal activity. Sesamol can also be used as an intermediate in the preparation of antidepressants such as Paroxetine (P205750). Sesamol has been found to be an antioxidant that may prevent the spoilage of oils. It also may prevent the spoilage of oils by acting as an antifungal. It is also used in ayurvedic Medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sesamol has good antioxidant activity. Sesamol has potential protective effects against free radicals as well as some antifungal activity. Sesamol can also be used as an intermediate in the preparation of antidepressants such as Paroxetine (P205750). Sesamol has been found to be an antioxidant that may prevent the spoilage of oils. It also may prevent the spoilage of oils by acting as an antifungal. It is also used in ayurvedic Medicine.

Solubility
Soluble in methanol, water (partly), and dichloromethane (25 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. N Erkan.; G Ayranci.; E Ayranci . Antioxidant activities of rosemary (Rosmarinus Officinalis L.) extract, blackseed (Nigella sativa L.) essential oil, carnosic acid, rosmarinic acid and sesamol. Food Chemistry. 2008110,(1) , 76-82.
  2. H Yoshida.; S Takagi. Antioxidative effects of sesamol and tocopherols at various concentrations in oils during microwave heating. Journal of the Science of Food and Agriculture. 199979,(2) , 220-226.