Thermo Scientific Chemicals

Sesamol, 98%, Thermo Scientific Chemicals

Catalog number: A10453.18
50 g, Each
Thermo Scientific Chemicals

Sesamol, 98%, Thermo Scientific Chemicals

Catalog number: A10453.18
50 g, Each
Quantity
Catalog number: A10453.18
also known as A10453-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
533-31-3
IUPAC Name
2H-1,3-benzodioxol-5-ol
Molecular Formula
C7H6O3
InChI Key
LUSZGTFNYDARNI-UHFFFAOYSA-N
SMILES
OC1=CC=C2OCOC2=C1
Melting Point (clear melt)
61-66?C
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
White to brown
Assay (GC)
≥97.5%

Description

Sesamol has good antioxidant activity. Sesamol has potential protective effects against free radicals as well as some antifungal activity. Sesamol can also be used as an intermediate in the preparation of antidepressants such as Paroxetine (P205750). Sesamol has been found to be an antioxidant that may prevent the spoilage of oils. It also may prevent the spoilage of oils by acting as an antifungal. It is also used in ayurvedic Medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sesamol has good antioxidant activity. Sesamol has potential protective effects against free radicals as well as some antifungal activity. Sesamol can also be used as an intermediate in the preparation of antidepressants such as Paroxetine (P205750). Sesamol has been found to be an antioxidant that may prevent the spoilage of oils. It also may prevent the spoilage of oils by acting as an antifungal. It is also used in ayurvedic Medicine.

Solubility
Soluble in methanol, water (partly), and dichloromethane (25 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text