Thermo Scientific Chemicals

Dibromomethane, 99%, Thermo Scientific Chemicals

Catalog number: A10456.0E
2500 g, Each
Thermo Scientific Chemicals

Dibromomethane, 99%, Thermo Scientific Chemicals

Catalog number: A10456.0E
2500 g, Each
Quantity
Catalog number: A10456.0E
also known as A10456-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
74-95-3
IUPAC Name
dibromomethane
Molecular Formula
CH2Br2
InChI Key
FJBFPHVGVWTDIP-UHFFFAOYSA-N
SMILES
BrCBr
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.5%
Refractive Index
1.5395-1.5425 @ 20?C

Description

Dibromomethane is used as solvent in organic synthesis. It acts as an intermediate in the manufacture of specialty chemicals, agrochemicals and pharmaceuticals. It is useful as extractant and utilized for the determination of 5-nitroimidazoles (5-NDZ) in environmental waters. It is involved in the convertion of catechols to their methylenedioxy derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibromomethane is used as solvent in organic synthesis. It acts as an intermediate in the manufacture of specialty chemicals, agrochemicals and pharmaceuticals. It is useful as extractant and utilized for the determination of 5-nitroimidazoles (5-NDZ) in environmental waters. It is involved in the convertion of catechols to their methylenedioxy derivatives.

Solubility
Miscible with water, chloroform, acetone, ether and alcohol.

Notes
Store in cool place. Incompatible with strong oxidizing agents, aluminum and magnesium.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text