Thermo Scientific Chemicals

Diethylammonium diethyldithiocarbamate, 98%, Thermo Scientific Chemicals

Catalog number: A10458.06
5 g, Each
Thermo Scientific Chemicals

Diethylammonium diethyldithiocarbamate, 98%, Thermo Scientific Chemicals

Catalog number: A10458.06
5 g, Each
Quantity
Catalog number: A10458.06
also known as A10458-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1518-58-7
IUPAC Name
diethyl[sulfanyl(carbonothioyl)]amine; diethylamine
Molecular Formula
C9H22N2S2
InChI Key
NBGTWXBPCIHUQD-UHFFFAOYSA-N
SMILES
CCNCC.CCN(CC)C(S)=S
Water Content (Karl Fischer Titration)
≤1%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
79.0-85.0?C
Appearance (Color)
White
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

Diethylammonium diethyldithiocarbamate is used in the synthesis of Dithiocarbamate. It serve as hosts for the specific binding of various metal cations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylammonium diethyldithiocarbamate is used in the synthesis of Dithiocarbamate. It serve as hosts for the specific binding of various metal cations.

Solubility
Soluble in Chloroform, Methanol.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Incompatible with strong acids, Strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text