Thermo Scientific Chemicals

1-Bromopropane, 99%, Thermo Scientific Chemicals

Catalog number: A10461.AK
250 mL, Each
Thermo Scientific Chemicals

1-Bromopropane, 99%, Thermo Scientific Chemicals

Catalog number: A10461.AK
250 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A10461.AK
also known as A10461-AK
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
106-94-5
IUPAC Name
1-bromopropane
Molecular Formula
C3H7Br
InChI Key
CYNYIHKIEHGYOZ-UHFFFAOYSA-N
SMILES
CCCBr
Appearance (Color)
Clear colorless to pale yellow
Comment
May darken on storage
Identification (FTIR)
Conforms (non-U.S. specification)
Water Content (Karl Fischer Titration)
≤0.01% (100 ppm) (U.S. specification)
Refractive Index
1.4325-1.4360 @ 20°C (non-U.S. specification)

Description

1-Bromopropane is used as a solvent for adhesives, degreasing plastics and optics. It is used in the production of asphalt, synthetic fiber and aerosols. It is employed for cleaning the metal surfaces, electronic circuit boards, and multi-layered OLEDs (Organic Light Emitting Diodes). It serves to replace perchloroethylene as a dry cleaning solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromopropane is used as a solvent for adhesives, degreasing plastics and optics. It is used in the production of asphalt, synthetic fiber and aerosols. It is employed for cleaning the metal surfaces, electronic circuit boards, and multi-layered OLEDs (Organic Light Emitting Diodes). It serves to replace perchloroethylene as a dry cleaning solvent.

Solubility
Miscible with ethanol, water and diethyl ether.

Notes
Light sensitive. Keep container tightly closed in a dry and well-ventilated place.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text