Thermo Scientific Chemicals

Sodium D-gluconate, 97%, Thermo Scientific Chemicals

Catalog number: A10464.0C
10 kg, Each
Thermo Scientific Chemicals

Sodium D-gluconate, 97%, Thermo Scientific Chemicals

Catalog number: A10464.0C
10 kg, Each
Quantity
Catalog number: A10464.0C
also known as A10464-0C
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
527-07-1
IUPAC Name
sodium 2,3,4,5,6-pentahydroxyhexanoate
Molecular Formula
C6H11NaO7
InChI Key
UPMFZISCCZSDND-UHFFFAOYNA-M
SMILES
[Na+].OCC(O)C(O)C(O)C(O)C([O-])=O
Appearance (Color)
White
Assay from Suppliers CofA
≥96.0%
Form
Powder or crystals or crystalline powder

Description

Sodium D-gluconate is a good chelator at alkaline pH, with chelating power relatively better than EDTA, NTA and other chelators. It has wide range of industrial applications which include textile dyeing, water treatments, printing and as cleaning agent for glass bottles. It also finds use as derusting agent in metallurgy and as additive in cement. It is used in food stuffs as bitterness inhibitor and as electrolyte replenisher.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium D-gluconate is a good chelator at alkaline pH, with chelating power relatively better than EDTA, NTA and other chelators. It has wide range of industrial applications which include textile dyeing, water treatments, printing and as cleaning agent for glass bottles. It also finds use as derusting agent in metallurgy and as additive in cement. It is used in food stuffs as bitterness inhibitor and as electrolyte replenisher.

Solubility
Very soluble in water; sparingly soluble in alcohol; insoluble in ether.

Notes
In general it is stable, non corrosive, non-toxic, readily biodegradable and incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text