Gateway™ pENTR™ 11 Dual Selection Vector
Product Image
Invitrogen™

Gateway™ pENTR™ 11 Dual Selection Vector

Gateway™ entry vectors are designed to clone DNA sequences using restriction endonucleases and ligase to create a Gateway™ entry clone.Read more
Have Questions?
Catalog number A10467
Price (USD)/ 10 µg
279.00
-
Add to cart
Price (USD)/ 10 µg
279.00
Add to cart
Gateway™ pENTR™ 11 Dual Selection Vector
Catalog numberA10467
Price (USD)/ 10 µg
279.00
-
Add to cart
Gateway™ entry vectors are designed to clone DNA sequences using restriction endonucleases and ligase to create a Gateway™ entry clone. The resulting entry clone is ready for recombination with a destination vector to create an expression clone. New: pENTR™ Dual Selection vectors!

The Gateway™ entry vectors (Table 1) offer the following:
• attL1 and attL2 sites for site-specific recombination of the entry clone with a Gateway™ destination vector to ensure cloning of the gene of interest in the correct orientation for expression
• Kozak consensus sequence for efficient translation initiation in eukaryotic systems
• Ribosome binding site for efficient translation initiation in prokaryotic systems (pENTR™ 1A Dual Selection, pENTR™3C Dual Selection, and pENTR™11 Dual Selection vectors only)
• rrnB transcription termination sequences to prevent basal expression of the PCR product of interest in E. coli
• pUC origin for high-copy replication and maintenance of the plasmid in E. coli
• Kanamycin resistance gene for selection in E. coli
• The ccdB⁄chloramphenicol fusion gene located between the two attL sites for
o negative selection and
o Chloramphenicol selection in E. coli
• Kanamycin resistance gene for selection in E. coli
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeDual SelectionExpression Vector
Antibiotic Resistance BacterialChloramphenicol (CmR), Kanamycin (KanR)
CleavageNo Cleavage Site
Protein TagUntagged
Cloning MethodGateway
Quantity10 μg
VectorpENTR
Product LinepENTR™
Unit Size10 µg
Contents & Storage
10 μg pENTR™ Dual Selection vector, in 20 ul in TE buffer, pH 8.0.
Store at -20°C