Thermo Scientific Chemicals

1,3,5-Trimethoxy-2-nitrobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A10473.06
5 g, Each
Thermo Scientific Chemicals

1,3,5-Trimethoxy-2-nitrobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A10473.06
5 g, Each
Quantity
Catalog number: A10473.06
also known as A10473-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14227-18-0
IUPAC Name
1,3,5-trimethoxy-2-nitrobenzene
Molecular Formula
C9H11NO5
InChI Key
VWYAWLZEMLQGJH-UHFFFAOYSA-N
SMILES
COC1=CC(OC)=C(C(OC)=C1)[N+]([O-])=O
Melting Point (clear melt)
149-155?C
Assay (GC)
≥97.5%
Form
Crystalline powder
Appearance (Color)
Yellow to orange

Description

2,4,6-Trimethoxynitrobenzene is used as a chemicals, pharmaceutical intermediate, chemical research, application intermediate, pharmaceutical reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethoxynitrobenzene is used as a chemicals, pharmaceutical intermediate, chemical research, application intermediate, pharmaceutical reagents.

Solubility
Soluble in water.

Notes
Desiccate at room temperature. Incompatabile to strong oxidizing agents, strong acids and bases. Store in a cool, dry, well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text