Thermo Scientific Chemicals

2,6-Lutidine, 98+%, Thermo Scientific Chemicals

Catalog number: A10478.AC
25 mL, Each
Thermo Scientific Chemicals

2,6-Lutidine, 98+%, Thermo Scientific Chemicals

Catalog number: A10478.AC
25 mL, Each
Quantity
Catalog number: A10478.AC
also known as A10478-AC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-48-5
IUPAC Name
2,6-dimethylpyridine
Molecular Formula
C7H9N
InChI Key
OISVCGZHLKNMSJ-UHFFFAOYSA-N
SMILES
CC1=CC=CC(C)=N1
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥98.0%
Water Content (Karl Fischer Titration)
<0.5%
Comment
May darken on storage
Refractive Index
1.4950-1.4990 @ 20?C

Description

2,6-Lutidine is used as a solvent in organic synthesis and as a sterically hindered mild base. It is also used as a vulcanization accelerator for dyes, resins and rubber. It also acts as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Lutidine is used as a solvent in organic synthesis and as a sterically hindered mild base. It is also used as a vulcanization accelerator for dyes, resins and rubber. It also acts as a food additive.

Solubility
Miscible with water.

Notes
Incompatible with acids, acid chlorides, oxidizing agents and chloroformates.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text