Thermo Scientific Chemicals

3-Hydroxy-4-methoxycinnamic acid, predominantly trans, 98+%, Thermo Scientific Chemicals

Catalog number: A10482.03
1 g, Each
Thermo Scientific Chemicals

3-Hydroxy-4-methoxycinnamic acid, predominantly trans, 98+%, Thermo Scientific Chemicals

Catalog number: A10482.03
1 g, Each
Quantity
Catalog number: A10482.03
also known as A10482-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
537-73-5
IUPAC Name
(2E)-3-(3-hydroxy-4-methoxyphenyl)prop-2-enoic acid
Molecular Formula
C10H10O4
InChI Key
QURCVMIEKCOAJU-HWKANZROSA-N
SMILES
COC1=CC=C(C=CC(O)=O)C=C1O
Assay (HPLC)
≥98.0%
Identification (FTIR)
Conforms
Appearance (Color)
White to pale cream
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Form
Crystals or powder or crystalline powder

Description

3-Hydroxy-4-methoxycinnamic acid exhibits hypoglycemic properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxy-4-methoxycinnamic acid exhibits hypoglycemic properties.

Solubility
Soluble in ethanol and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents. Stable under normal temperatures and pressures.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text