Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Catalog Number | Quantity |
---|---|
A1048901 | 500 mL |
Minimum Essential Medium (MEM) is one of the most commonly used of all cell culture media. MEM can be used with a variety of suspension and adherent mammalian cells, including HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, fibroblasts, and primary rat astrocytes. We offer a variety of Gibco™ MEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
With | Without |
L-glutamine | Gentamycin |
Proline | HEPES |
Sodium Pyruvate | Insulin |
Zinc | |
Phenol Red |
The complete formulation is available.
Gibco™ Richter's MEM is a modified formulation of Eagle's MEM that contains iron, zinc, putrescine, fatty acids, and gentamycin. Richter's modification also includes very high concentrations of both choline chloride and i-Inositol. This product was further modified with L-glutamine, and without gentamicin sulfate.
Product Use
For Research Use Only. Not for use in diagnostic procedures.
Gibco™ MEM contains no proteins, lipids, or growth factors. Therefore, MEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Share catalog number, name or link