Diphenylsilanediol, 98%, Thermo Scientific Chemicals
Diphenylsilanediol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diphenylsilanediol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A10492.22
also known as A10492-22
Price (USD)/ Each
81.10
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
81.10
Add to cart
Diphenylsilanediol, 98%, Thermo Scientific Chemicals
Catalog numberA10492.22
Price (USD)/ Each
81.10
-
Add to cart
Chemical Identifiers
CAS947-42-2
IUPAC Namediphenylsilanediol
Molecular FormulaC12H12O2Si
InChI KeyOLLFKUHHDPMQFR-UHFFFAOYSA-N
SMILESO[Si](O)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)≥97.5%
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale cream to pale gray
Identification (FTIR)Conforms
Diphenylsilanediol is used in Medicine field, electronics industry and polymer material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylsilanediol is used in Medicine field, electronics industry and polymer material.

Solubility
Reacts with water.

Notes
Stable under normal temperatures and pressures. Incompatible with Oxidizing agents. Store at room temperature. Air & Light Sensitive.
RUO – Research Use Only
J. Keith Fawcett, Norman Camerman, Arthur Camerman. Stereochemical basis of anticonvulsant drug action. VI. Crystal and molecular structure of diphenylsilanediol. Canadian Journal of Chemistry.1977, 55, (20), 3631-3635.