SequalPrep™ Long PCR Kit with dNTPs
For long PCR with >100x Taq fidelity and 30 sec/kb synthesis, use Invitrogen Platinum SuperFi DNA Polymerase.
Product Image
Applied Biosystems™

SequalPrep™ Long PCR Kit with dNTPs

The SequalPrep™ Long PCR Kit with dNTPs provides the highest amplification success rate of any long range PCR kit testedRead more
Have Questions?
Catalog NumberQuantity
A104981,000 units
Catalog number A10498
Price (USD)
2,400.00
Each
Add to cart
Quantity:
1,000 units
Price (USD)
2,400.00
Each
Add to cart
The SequalPrep™ Long PCR Kit with dNTPs provides the highest amplification success rate of any long range PCR kit tested across multiple organisms and platforms for non-optimized amplicons up to 20 kB with little to no optimization. Successful long range PCR with minimal optimization enables efficient PCR enrichment for a variety of applications including medical re-sequencing on next generation sequencing (NGS) platforms.
PCR amplification is the traditional method to enrich target regions of the genome for genotyping and re-sequencing applications. To take advantage of the enhanced NGS processing capabilities, higher throughput enrichment methods have become necessary. Unfortunately, the potential efficiency gains from long range PCR using available commercial solutions have been offset with lower amplification success rates limiting enrichment to ∼5 kB regions. The SequalPrep™ Long PCR Kit with dNTPs overcomes these issues and provides the highest amplification success rate of any commercially available kit tested across multiple organisms and platforms for amplicons up to 20 kB with minimal optimization.

This kit can be used upstream of any NGS platform or for other genotyping applications. When combined with the SequalPrep™ Normalization Plate the SequalPrep™ (Sequence Equalization Sample Preparation) System is the first complete PCR enrichment and amplicon equalization solution that provides a viable and efficient enrichment method to meet NGS re-sequencing throughput capabilities. This NGS Sample Prep system significantly reduces PCR amplification and normalization complexity to allow for enrichment of regions up to 1MB in a 96 well plate format. Amplify and equalize with the ease of SequalPrep™ for consistently high coverage of your regions of interest.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Exonuclease Activity3' - 5', 5' - 3'
Fidelity (vs. Taq)4 X
Hot StartBuilt-In Hot Start
OverhangMixed
PolymeraseSequalPrep Long Polymerase
Product LineSequalPrep™
Quantity1,000 units
Reaction FormatSeparate Components
Shipping ConditionDry Ice
Size (Final Product)20 kb or less
GC-Rich PCR PerformanceMedium
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
• 200 μL SequalPrep™ Long Polymerase (5 U/μL)
• 1.2 ml SequalPrep™ 10X Reaction Buffer
• 1.2 mL SequalPrep™ 10X Enhancer A
• 1.2 mL SequalPrep™ 10X Enhancer B
• 750 μL DMSO

Store at -20°C. DMSO may be stored at 4°C or room temperature.

Frequently asked questions (FAQs)

What is the fidelity of the SequalPrep Long Polymerase in SequalPrep Long PCR Kit with dNTPs?

The SequalPrep Long PCR Kit with dNTPs demonstrates fidelity 3-4 times greater than recombinant Taq polymerase, similar to Invitrogen’s Platinum Taq High Fidelity Enzyme (by rpsl assay).  The SequalPrep Long PCR Polymerase was compared to rTaq, Platinum Taq HiFi, & Roche's Expand Long Template PCR System.  Relative fidelity:
• rTaq = 1
• Platinum Taq HiFi = 3.3
• Roche = 3.6
• SequalPrep Long with Enhancer A = 3.6
• SequalPrep Long with Enhancer B = 4.4

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

What organisms have been tested with SequalPrep Long PCR Kit with dNTPs kit?

Human and mouse genomic DNA have both been tested with the kit.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

SequalPrep Long PCR Kit with dNTPs comes with several enhancers. What are they and when should I use them?

The two Enhancers were selected to help provide maximum success rates. We recommend setting up the initial reactions with 0.5X Enhancer A to provide maximum success rate.  For any unsuccessful amplicons we recommend testing the following conditions to optimize amplification:
• 1.0X Enhancer A
• 0.5X Enhancer B
• 1.0X Enhancer B

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

How many units of the SequalPrep Long Polymerase should I use in a typical reaction for SequalPrep Long PCR Kit with dNTPs?

We recommend using 1.8 Units of SequalPrep Long Polymerase in a reaction.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

What amount of input material do you recommend for SequalPrep Long PCR Kit with dNTPs?

We recommend 5-200 ng of human genomic DNA and 20-80 ng of mouse embryonic stem cells DNA. 50 ng of DNA template at a concentration of 50 ng/µl should be sufficient for most applications, although input amounts as low as 10 ng can be possible depending on the sample type, due to the high yields generated by the kit.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link