Thermo Scientific Chemicals

Glutaraldehyde, 50% aq. soln., Thermo Scientific Chemicals

Catalog number: A10500.36
500 g, Each
Thermo Scientific Chemicals

Glutaraldehyde, 50% aq. soln., Thermo Scientific Chemicals

Catalog number: A10500.36
500 g, Each
Quantity
Catalog number: A10500.36
also known as A10500-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-30-8
IUPAC Name
pentanedial
Molecular Formula
C5H8O2
InChI Key
SXRSQZLOMIGNAQ-UHFFFAOYSA-N
SMILES
O=CCCCC=O
Form
Liquid
Comment
May contain up to 10% methanol
Appearance (Color)
Clear colorless to pale yellow
Assay (Titration)
≥48.0 to ≤52.0%

Description

Glutaraldehyde is used as a cross linking agent for gelatin, poly(vinyl alcohol) and polyheptapeptides. It is also used as a fixative for electron microscopy and as disinfectants. It provides water resistance to protein and polyhydroxy compounds by reacting through cross linking. It acts as an intermediate for the production of resins, dyestuffs, pharmaceuticals, photographic films and for stabilization of proteins on agarose beads, activation of polystyrene and glass for immobilization of antibodies and antigens and coupling peptides onto carrier proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glutaraldehyde is used as a cross linking agent for gelatin, poly(vinyl alcohol) and polyheptapeptides. It is also used as a fixative for electron microscopy and as disinfectants. It provides water resistance to protein and polyhydroxy compounds by reacting through cross linking. It acts as an intermediate for the production of resins, dyestuffs, pharmaceuticals, photographic films and for stabilization of proteins on agarose beads, activation of polystyrene and glass for immobilization of antibodies and antigens and coupling peptides onto carrier proteins.

Solubility
Miscible with water, acetone, dichloromethane, ethylacetate, isopropanol, n-hexane and toluene.

Notes
Incompatible with strong oxidisng agents, strong acids and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text