Thermo Scientific Chemicals

4-(Trifluoromethyl)benzyl alcohol, 98%, Thermo Scientific Chemicals

Catalog number: A10508.22
100 g, Each
Thermo Scientific Chemicals

4-(Trifluoromethyl)benzyl alcohol, 98%, Thermo Scientific Chemicals

Catalog number: A10508.22
100 g, Each
Quantity
Catalog number: A10508.22
also known as A10508-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
349-95-1
IUPAC Name
[4-(trifluoromethyl)phenyl]methanol
Molecular Formula
C8H7F3O
InChI Key
MOOUWXDQAUXZRG-UHFFFAOYSA-N
SMILES
OCC1=CC=C(C=C1)C(F)(F)F
Form
Liquid
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%

Description

4-(Trifluoromethyl)benzyl alcohol, is used as a pharmaceutical intermediate, it is also used to predict the NMR spectrum. 4-(Trifluoromethyl)benzyl alcohol is employed as a reagent in, for example, kinetic studies of phosphonoformate prodrugs and aquachromium(IV).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)benzyl alcohol, is used as a pharmaceutical intermediate, it is also used to predict the NMR spectrum. 4-(Trifluoromethyl)benzyl alcohol is employed as a reagent in, for example, kinetic studies of phosphonoformate prodrugs and aquachromium(IV).

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Keep away from oxidizing agents,reducing agents, acid chlorides, acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text