Thermo Scientific Chemicals

3,5-Di-tert-butyl-4-hydroxycinnamic acid, predominantly trans, 97%, Thermo Scientific Chemicals

Catalog number: A10514.09
10 g, Each
Thermo Scientific Chemicals

3,5-Di-tert-butyl-4-hydroxycinnamic acid, predominantly trans, 97%, Thermo Scientific Chemicals

Catalog number: A10514.09
10 g, Each
Quantity
Catalog number: A10514.09
also known as A10514-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22014-01-3
IUPAC Name
(2E)-3-(3,5-di-tert-butyl-4-hydroxyphenyl)prop-2-enoic acid
Molecular Formula
C17H24O3
InChI Key
CTYWXRDQWMRIIM-BQYQJAHWSA-N
SMILES
CC(C)(C)C1=CC(C=CC(O)=O)=CC(=C1O)C(C)(C)C
Appearance (Color)
Pale cream to cream to yellow to orange
Form
Powder
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Assay (Silylated GC)
≥96.0%

Description

3,5-Di-tert-butyl-4-hydroxycinnamic acid, predominantly trans,

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Di-tert-butyl-4-hydroxycinnamic acid, predominantly trans,

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text