1,4-Dibromobenzene, 98%, Thermo Scientific Chemicals
1,4-Dibromobenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,4-Dibromobenzene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
50 g
100 g
250 g
Catalog number A10517.0E
also known as A10517-0E
Price (USD)/ Each
327.65
Online exclusive
364.00 
Save 36.35 (10%)
-
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)/ Each
327.65
Online exclusive
364.00 
Save 36.35 (10%)
Add to cart
1,4-Dibromobenzene, 98%, Thermo Scientific Chemicals
Catalog numberA10517.0E
Price (USD)/ Each
327.65
Online exclusive
364.00 
Save 36.35 (10%)
-
Add to cart
Chemical Identifiers
CAS106-37-6
IUPAC Name1,4-dibromobenzene
Molecular FormulaC6H4Br2
InChI KeySWJPEBQEEAHIGZ-UHFFFAOYSA-N
SMILESBrC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)85.0-92.0?C
Appearance (Color)White
1,4-Dibromobenzene is used in the synthesis of 2,5-dibromoacetophenone, triarylamines and trans- stilbenes. It is widely used as a heavy liquid solvent as well as a motor oil additive. It finds application as an intermediate in the manufacture of organic chemicals such as dyes, pharmaceuticals and flame retardants for polymeric materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dibromobenzene is used in the synthesis of 2,5-dibromoacetophenone, triarylamines and trans- stilbenes. It is widely used as a heavy liquid solvent as well as a motor oil additive. It finds application as an intermediate in the manufacture of organic chemicals such as dyes, pharmaceuticals and flame retardants for polymeric materials.

Solubility
Soluble in alcohol, benzene, chloroform, toluene and carbon tetrachloride. Insoluble in ether.

Notes
Avoid excessive heat and light. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. In the presence of NiCl2, cross-coupling with aryl Grignards gives p-terphenyls: Synthesis, 147 (1990).
  2. Cano, N. H.; Ballari, M. S.; Lopez, A. G.; Santiago, A. N. New Synthesis and Biological Evaluation of Benzothiazole Derivates as Antifungal Agents. J. Agric. Food Chem. 2015, 63 (14), 3681-3686.
  3. Vazdar, M.; Mitchell, A. S.; Warrener, R. N.; Margetic, D. High pressure promoted exchange of guests from hemicarceplexes. Tetrahedron 2015, 71 (4), 550-553.