Thermo Scientific Chemicals

Glycylglycine, 99%

Catalog number: A10523.14
25 g, Each
Thermo Scientific Chemicals

Glycylglycine, 99%

Catalog number: A10523.14
25 g, Each
Quantity
Catalog number: A10523.14
also known as A10523-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
556-50-3
IUPAC Name
hydrogen 2-(2-aminoacetamido)acetic acid chloride
Molecular Formula
C4H9ClN2O3
InChI Key
YHBAZQDEMYQPJL-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NCC(=O)NCC(O)=O
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Appearance (Color)
White
Form
Crystals or powder or crystalline powder

Description

Glycylglycine is as a buffering reagent and also used in biochemical research. It is the simplest of all dipeptides and is used as a starting template for preparation of more complex peptides. It is also employed as a practical buffer with a buffering range of 7.5 - 8.9 (at 25°C). Valuable agent for experiments investigating peptide transport and absorption.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycylglycine is as a buffering reagent and also used in biochemical research. It is the simplest of all dipeptides and is used as a starting template for preparation of more complex peptides. It is also employed as a practical buffer with a buffering range of 7.5 - 8.9 (at 25°C). Valuable agent for experiments investigating peptide transport and absorption.

Solubility
Soluble in water (132 g/L) at 20°C, and ethanol (slightly solube).

Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text