5-Nitrobenzimidazole is used as sensitive reagents determinating of bismuth, cadmium, copper,and rubber antioxidant. It also used as photographic materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Nitrobenzimidazole is used as sensitive reagents determinating of bismuth, cadmium, copper,and rubber antioxidant. It also used as photographic materials.
Solubility
Sparingly soluble in water. (0.5g/L at 25°C). Soluble in methanol.
Notes
Incompatible with strong oxidizing agents, strong bases. Store at room temperature.
RUO – Research Use Only
Dana Deylova; Bogdan Yosypchuk; Vlastimil Vyskocil; Jiri Barek. Voltammetric Determination of 4-Nitrophenol and 5-Nitrobenzimidazole Using Different Types of Silver Solid Amalgam Electrodes - A Comparative Study. Electroanalysis. 2011, 23, (7),1548-1555
Ayse E.Puetuen; Goezen Bereket; Esra Keskin. Potentiometric Titrations of Some 2-Substituted 5-Nitrobenzimidazole Derivatives in Nonaqueous Solvent. J. Chem. Eng. Data. 1995, 40, (1),221-224