Valerophenone, 99%, Thermo Scientific Chemicals
Valerophenone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Valerophenone, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A10525.14
also known as A10525-14
Price (USD)/ Each
41.65
Online exclusive
46.60 
Save 4.95 (11%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
41.65
Online exclusive
46.60 
Save 4.95 (11%)
Add to cart
Valerophenone, 99%, Thermo Scientific Chemicals
Catalog numberA10525.14
Price (USD)/ Each
41.65
Online exclusive
46.60 
Save 4.95 (11%)
-
Add to cart
Chemical Identifiers
CAS1009-14-9
IUPAC Name1-phenylpentan-1-one
Molecular FormulaC11H14O
InChI KeyXKGLSKVNOSHTAD-UHFFFAOYSA-N
SMILESCCCCC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colourless to pale yellow
Assay (GC)>98.5%
Formliquid
Refractive Index1.5130-1.5160
Valerophenone is used as intermediates of liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Valerophenone is used as intermediates of liquid crystals.

Solubility
Insoluble in water.

Notes
 Incompatible with strong oxidizing agents, acids, bases, plastics. Store at room temperature with proper ventilation.
RUO – Research Use Only

General References:

  1. R.G.Zepp; M.M.Gumz; W.L.Miller; H.Gao. Photoreaction of Valerophenone in Aqueous Solution. J. Phys. Chem. A. 1998, 102, (28),5716-5723
  2. P.Novak; J.Matousek; J.Briza. Valerophenone synthase-like chalcone synthase homologues in Humulus lupulus. Biologia Plantarum. 2003, 46, (3),375-381