Thermo Scientific Chemicals

Valerophenone, 99%, Thermo Scientific Chemicals

Catalog number: A10525.36
500 g, Each
Thermo Scientific Chemicals

Valerophenone, 99%, Thermo Scientific Chemicals

Catalog number: A10525.36
500 g, Each
Quantity
Catalog number: A10525.36
also known as A10525-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1009-14-9
IUPAC Name
1-phenylpentan-1-one
Molecular Formula
C11H14O
InChI Key
XKGLSKVNOSHTAD-UHFFFAOYSA-N
SMILES
CCCCC(=O)C1=CC=CC=C1
Appearance (Color)
Clear, colourless to pale yellow
Assay (GC)
>98.5%
Form
liquid
Refractive Index
1.5130-1.5160

Description

Valerophenone is used as intermediates of liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Valerophenone is used as intermediates of liquid crystals.

Solubility
Insoluble in water.

Notes
 Incompatible with strong oxidizing agents, acids, bases, plastics. Store at room temperature with proper ventilation.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text