Thermo Scientific Chemicals

Ammonium peroxydisulfate, 98%, Thermo Scientific Chemicals

Catalog number: A10533.36
500 g, Each
Thermo Scientific Chemicals

Ammonium peroxydisulfate, 98%, Thermo Scientific Chemicals

Catalog number: A10533.36
500 g, Each
Quantity
Catalog number: A10533.36
also known as A10533-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7727-54-0
IUPAC Name
diammonium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular Formula
H8N2O8S2
InChI Key
ROOXNKNUYICQNP-UHFFFAOYSA-N
SMILES
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O
Appearance (Color)
White to pale yellow
Assay from Suppliers CofA
≥97.5% (U.S. specification)
Form
Crystalline powder or crystals
Comment
Aqueous solution decomposes slowly at room temperature.
Impurity content
Iron (Fe): ≤10 ppm (non-U.S. specification)

Description

Ammonium peroxydisulfate is widely utilized as an oxidizing agent and radical initiators in the polymerization of certain alkenes. It is actively involved in in the synthesis of commercially important polymers like styrene-butadiene rubber and polytetrafluoroethylene. It combines with tetramethylethylenediamine to catalyze the polymerization of acrylamide. Due to its oxidizing properties, it can be employed to etch copper on printed circuit boards which is an alternative to ferric chloride solution. Also, it acts as oxidants in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium peroxydisulfate is widely utilized as an oxidizing agent and radical initiators in the polymerization of certain alkenes. It is actively involved in in the synthesis of commercially important polymers like styrene-butadiene rubber and polytetrafluoroethylene. It combines with tetramethylethylenediamine to catalyze the polymerization of acrylamide. Due to its oxidizing properties, it can be employed to etch copper on printed circuit boards which is an alternative to ferric chloride solution. Also, it acts as oxidants in organic synthesis.

Solubility
Freely soluble in water.

Notes
Aqueous solution decomposes slowly at room temperature.Moisture sensitive. Incompatible with strong reducing agents, organic materials and powdered metals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text