Thermo Scientific Chemicals

4,6-Dihydroxy-2-methylpyrimidine, 99%, Thermo Scientific Chemicals

Catalog number: A10535.09
10 g, Each
Thermo Scientific Chemicals

4,6-Dihydroxy-2-methylpyrimidine, 99%, Thermo Scientific Chemicals

Catalog number: A10535.09
10 g, Each
Quantity
Catalog number: A10535.09
also known as A10535-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
40497-30-1
Appearance (Color)
White to pale cream or pale pink
Identification (FTIR)
Conforms
Assay (Silylated GC)
≥98.5%
Form
Powder

Description

4,6-Dihydroxy-2-methylpyrimidine is used in the preparation of 4,6-dichloro-2-methyl-pyrimidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,6-Dihydroxy-2-methylpyrimidine is used in the preparation of 4,6-dichloro-2-methyl-pyrimidine.

Solubility
Soluble in sodium hydroxide.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text