Molybdic acid, MoO3 85% min, Thermo Scientific Chemicals
Molybdic acid, MoO3 85% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molybdic acid, MoO3 85% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2500 g
Catalog number A10537.36
also known as A10537-36
Price (USD)/ Each
120.65
Online exclusive
134.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
120.65
Online exclusive
134.00 
Save 13.35 (10%)
Add to cart
Molybdic acid, MoO3 85% min, Thermo Scientific Chemicals
Catalog numberA10537.36
Price (USD)/ Each
120.65
Online exclusive
134.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7782-91-4
IUPAC Namedioxomolybdenumdiol
Molecular FormulaH2MoO4
InChI KeyVLAPMBHFAWRUQP-UHFFFAOYSA-L
SMILESO[Mo](O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Elemental AnalysisChloride content (as Cl) ≤0.01%
Appearance (Color)White to pale cream
Assay (unspecified)MoO3 content >= 85.0 to =< 90.0%
FormPowder
Elemental AnalysisLead content (as Pb) ≤0.003%
Molybdic acid is used as a precursor in the production of ammonium molybdate tetrahydrate and molybdenum(IV) sulfide. The mixture of molybdic acid in concentrated sulfuric acid is called as Froehde reagent, which finds application in analytical chemistry for the detection of alkaloids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Molybdic acid is used as a precursor in the production of ammonium molybdate tetrahydrate and molybdenum(IV) sulfide. The mixture of molybdic acid in concentrated sulfuric acid is called as Froehde reagent, which finds application in analytical chemistry for the detection of alkaloids.

Solubility
Soluble in dilute ammonium hydroxide, alkali and ammonia. Slightly soluble in water.

Notes
Incompatible with strong bases and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Ahmed, N.; Siddiqui, Z. N. Silica molybdic acid catalysed eco-friendly three component synthesis of functionalised tetrazole derivatives under microwave irradiation in water. RSC Adv. 2015, 5 (22), 16707-16717.
  2. Zhou, J.; Lin, N.; Wang, L.; Zhang, K.; Zhu, Y.; Qian, Y. Synthesis of hexagonal MoO3 nanorods and a study of their electrochemical performance as anode materials for lithium-ion batteries. J. Mater. Chem. A 2015, 3 (14), 7463-7468.