2,6-Dichlorobenzoic acid, 98%, Thermo Scientific Chemicals
2,6-Dichlorobenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Dichlorobenzoic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A10542.14
also known as A10542-14
Price (USD)/ Each
56.20
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
56.20
Add to cart
2,6-Dichlorobenzoic acid, 98%, Thermo Scientific Chemicals
Catalog numberA10542.14
Price (USD)/ Each
56.20
-
Add to cart
Chemical Identifiers
CAS50-30-6
IUPAC Name2,6-dichlorobenzoic acid
Molecular FormulaC7H4Cl2O2
InChI KeyMRUDNSFOFOQZDA-UHFFFAOYSA-N
SMILESOC(=O)C1=C(Cl)C=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (Aqueous acid-base Titration)≥96.0% to ≤103.0%
Melting Point (clear melt)138.0-147.0?C
Appearance (Color)White to cream
Assay (Silylated GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only