Thermo Scientific Chemicals

4-Methoxytrityl chloride, 97%, Thermo Scientific Chemicals

Catalog number: A10545.30
250 g, Each
Thermo Scientific Chemicals

4-Methoxytrityl chloride, 97%, Thermo Scientific Chemicals

Catalog number: A10545.30
250 g, Each
Quantity
Catalog number: A10545.30
also known as A10545-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14470-28-1
IUPAC Name
1-(chlorodiphenylmethyl)-4-methoxybenzene
Molecular Formula
C20H17ClO
InChI Key
OBOHMJWDFPBPKD-UHFFFAOYSA-N
SMILES
COC1=CC=C(C=C1)C(Cl)(C1=CC=CC=C1)C1=CC=CC=C1
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
White to cream to pale orange or pink
Melting Point (clear melt)
118.0-127.0?C
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%

Description

It is used as selective protecting reagent that is used for primary hydroxyl groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as selective protecting reagent that is used for primary hydroxyl groups.

Solubility
Soluble in toluene.

Notes
It is hygroscopic. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text