Thermo Scientific Chemicals

Indole-3-acetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A10556.14
25 g, Each
Thermo Scientific Chemicals

Indole-3-acetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A10556.14
25 g, Each
Quantity
Catalog number: A10556.14
also known as A10556-14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
87-51-4
IUPAC Name
2-(1H-indol-3-yl)acetic acid
Molecular Formula
C10H9NO2
InChI Key
SEOVTRFCIGRIMH-UHFFFAOYSA-N
SMILES
OC(=O)CC1=CNC2=CC=CC=C12
Identification (FTIR)
Conforms
Appearance (Color)
Pale cream to cream to pale brown or pale pink to gray
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (Silylated GC)
≥97.5%

Description

It is applied as plant growth hormone. is an inducer of plant cell elongation and division shown to cause uncontrolled growth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as plant growth hormone. is an inducer of plant cell elongation and division shown to cause uncontrolled growth.

Solubility
Soluble in ethanol (50 mg/ml), methanol, DMSO, and chloroform (sparingly). Insoluble in water.

Notes
Light sensitive. Store at -20°C. ncompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text