Thermo Scientific Chemicals

N,N-Diethylaniline, 99%, Thermo Scientific Chemicals

Catalog number: A10560.AP
500 mL, Each
Thermo Scientific Chemicals

N,N-Diethylaniline, 99%, Thermo Scientific Chemicals

Catalog number: A10560.AP
500 mL, Each
Quantity
Catalog number: A10560.AP
also known as A10560-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
91-66-7
IUPAC Name
N,N-diethylaniline
Molecular Formula
C10H15N
InChI Key
GGSUCNLOZRCGPQ-UHFFFAOYSA-N
SMILES
CCN(CC)C1=CC=CC=C1
Appearance (Color)
Clear pale yellow to yellow
Assay (GC)
≥98.5%
Refractive Index
1.5405-1.5435 @ 20?C
Identification (FTIR)
Conforms
Form
Liquid

Description

N,N-Diethylaniline is used as an intermediate in pharmaceuticals, dyes and other chemicals. It acts as a reaction catalyst. Its complex, diethylaniline borane is used as a reducing agent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Diethylaniline is used as an intermediate in pharmaceuticals, dyes and other chemicals. It acts as a reaction catalyst. Its complex, diethylaniline borane is used as a reducing agent in organic synthesis.

Solubility
Miscible with water and acetone. Slightly miscible with chloroform, alcohol and ether.      

Notes
Incompatible with strong oxidizing agents and strong acids.                  .
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text