N,N,2,4,6-Pentamethylaniline, 98%, Thermo Scientific Chemicals
N,N,2,4,6-Pentamethylaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N,2,4,6-Pentamethylaniline, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
Catalog number A10561.14
also known as A10561-14
Price (USD)/ Each
91.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
91.00
Add to cart
N,N,2,4,6-Pentamethylaniline, 98%, Thermo Scientific Chemicals
Catalog numberA10561.14
Price (USD)/ Each
91.00
-
Add to cart
Chemical Identifiers
CAS13021-15-3
IUPAC NameN,N,2,4,6-pentamethylaniline
Molecular FormulaC11H17N
InChI KeyJZBZLRKFJWQZHU-UHFFFAOYSA-N
SMILESCN(C)C1=C(C)C=C(C)C=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless
Assay (GC)>97.5%
FormLiquid
Refractive Index1.5100-1.5140 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only