Trichloroacetonitrile, 98%, Thermo Scientific Chemicals
Trichloroacetonitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trichloroacetonitrile, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1000 g
Catalog number A10565.30
also known as A10565-30
Price (USD)/ Each
90.65
Online exclusive
101.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
90.65
Online exclusive
101.00 
Save 10.35 (10%)
Add to cart
Trichloroacetonitrile, 98%, Thermo Scientific Chemicals
Catalog numberA10565.30
Price (USD)/ Each
90.65
Online exclusive
101.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS545-06-2
IUPAC Nametrichloroacetonitrile
Molecular FormulaC2Cl3N
InChI KeyDRUIESSIVFYOMK-UHFFFAOYSA-N
SMILESClC(Cl)(Cl)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Refractive Index1.4390-1.4430 @ 20?C
Trichloroacetonitrile is involved as a reagent in Overman rearrangement, which is used to prepare alylic amines from allylic alcohols. It is also used to prepare bistrichloroacetimidates from diols leading to dihyrooxazines through acid catalyzed cyclization. Further, it is utilized in the synthesis of trichloroacetimidates by 1,8-Diazobicyclo[5.4.0]undec-7-ene (DBU) catalyzed addition of allylic alcohols. It finds application in the study of the methoxy methyl (MOM) catalyzed aza-Claisen rearrangement.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trichloroacetonitrile is involved as a reagent in Overman rearrangement, which is used to prepare alylic amines from allylic alcohols. It is also used to prepare bistrichloroacetimidates from diols leading to dihyrooxazines through acid catalyzed cyclization. Further, it is utilized in the synthesis of trichloroacetimidates by 1,8-Diazobicyclo[5.4.0]undec-7-ene (DBU) catalyzed addition of allylic alcohols. It finds application in the study of the methoxy methyl (MOM) catalyzed aza-Claisen rearrangement.

Solubility
Miscible with most organic solvents. Immiscible with water.

Notes
Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
RUO – Research Use Only

General References:

  1. The combination with PPh3 is superior to the popular CCl4-PPh3 system for conversion of carboxylic acids to acyl chlorides: Tetrahedron Lett., 40, 5323 (1999). It also converts allylic alcohols to the corresponding chlorides: Russ. J. Org. Chem., 31, 1019 (1995).
  2. In the presence of a base, e.g DBU, reacts with alcohols to form trichloroacetimidates, which are a useful means of protection, readily cleaved by TsOH/MeOH, DBU/MeOH or ZnH4Cl: Synlett, 753(1999). These imidates are activated towards displacement by nucleophiles. Glycosyl imidates, formed in the presence of a base such as K2CO3 or DBU, are widely applied as glycosyl donors in oligosaccharide synthesis. Reaction with the free OH of the glycosyl acceptor occurs under mild conditions, promoted by a Lewis acid, e.g. BF3 etherate or TMS-OTf, to form, respectively, either the ß- or ɑ-glycoside as the major product: Angew. Chem. Int. Ed., 25, 212 (1986). For reviews, see: Chem. Rev., 93, 1503 (1993); Adv. Carbohydr. Chem. Biochem., 50, 21 (1994); Contemp. Org. Synth., 3, 173 (1996). The imidates of allylic alcohols, prepared using NaH, or, more conveniently, KOH under phase-transfer conditions: Tetrahedron Lett., 37, 1481 (1996), undergo a [3,3]-sigmatropic rearrangement to derivatives of allylamines: J. Am. Chem. Soc., 96, 597 (1974); 98, 2901 (1976). For an example, see: Org. Synth. Coll., 6, 507 (1988). Review: Acc. Chem. Res., 13, 218 (1980):
  3. Aromatic aldoximes are dehydrated to nitriles by a similar cyclic mechanism: J. Org. Chem., 38, 2241 (1973).
  4. Reacts with hydrogen peroxide, to give, in situ, peroxytrichloroacetamidic acid, a reagent for the epoxidation of alkenes under essentially neutral conditions: J. Org. Chem., 48, 888 (1983).
  5. For a review of the chemistry of trichloroacetonitrile, see: Heterocycles, 43, 1083 (1996).