Thermo Scientific Chemicals

Dimethyl aminoterephthalate, 99%, Thermo Scientific Chemicals

Catalog number: A10567.18
50 g, Each
Thermo Scientific Chemicals

Dimethyl aminoterephthalate, 99%, Thermo Scientific Chemicals

Catalog number: A10567.18
50 g, Each
Quantity
Catalog number: A10567.18
also known as A10567-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5372-81-6
IUPAC Name
1,4-dimethyl 2-aminobenzene-1,4-dicarboxylate
Molecular Formula
C10H11NO4
InChI Key
DSSKDXUDARIMTR-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=C(C(=O)OC)C(N)=C1
Appearance (Color)
Cream to yellow to green to brown
Form
Powder
Melting Point (clear melt)
129.0-137.0?C
Assay (GC)
≥98.5%

Description

Dimethyl aminoterephthalate is used in the preparation of imidazotetrazinones as probes for the action of temozolomide. It is also used to prepare aminoquinazolines with neurokinin-2 receptor antagonist activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl aminoterephthalate is used in the preparation of imidazotetrazinones as probes for the action of temozolomide. It is also used to prepare aminoquinazolines with neurokinin-2 receptor antagonist activity.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text