Thermo Scientific Chemicals

D-Phenylalanine, 99%

Catalog number: A10572.06
5 g, Each
Thermo Scientific Chemicals

D-Phenylalanine, 99%

Catalog number: A10572.06
5 g, Each
Quantity
Catalog number: A10572.06
also known as A10572-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
673-06-3
IUPAC Name
(2R)-2-amino-3-phenylpropanoic acid
Molecular Formula
C9H11NO2
InChI Key
COLNVLDHVKWLRT-MRVPVSSYSA-N
SMILES
N[C@H](CC1=CC=CC=C1)C(O)=O
Appearance (Color)
White to pale cream
Optical Rotation
34.0 ± 2.0? (C=2 in water)
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms

Description

It is an essential amino acid with anti-depressant and analgesic activities. pharmaceutical intermediates, mainly used to treat, prevent osteoporosis, cardiovascular, diabetes, arterioscleros.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an essential amino acid with anti-depressant and analgesic activities. pharmaceutical intermediates, mainly used to treat, prevent osteoporosis, cardiovascular, diabetes, arterioscleros.

Solubility
Soluble in water (30 mg/ml). Slightly soluble in methanol, ethanol, DMSO.

Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text