Thermo Scientific Chemicals

3-Nitrobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A10579.30
250 g, Each
Thermo Scientific Chemicals

3-Nitrobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A10579.30
250 g, Each
Quantity
Catalog number: A10579.30
also known as A10579-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
121-90-4
IUPAC Name
3-nitrobenzoyl chloride
Molecular Formula
C7H4ClNO3
InChI Key
NXTNASSYJUXJDV-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=CC(=C1)C(Cl)=O
Form
Fused solid
Melting Point (clear melt)
29.0-36.0?C
Identification (FTIR)
Conforms
Appearance (Color)
White to pale yellow to pale brown
Assay (GC)
≥97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text