Thermo Scientific Chemicals

Deoxybenzoin, 97%, Thermo Scientific Chemicals

Catalog number: A10580.22
100 g, Each
Thermo Scientific Chemicals

Deoxybenzoin, 97%, Thermo Scientific Chemicals

Catalog number: A10580.22
100 g, Each
Quantity
Catalog number: A10580.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
451-40-1
IUPAC Name
1,2-diphenylethan-1-one
Molecular Formula
C14H12O
InChI Key
OTKCEEWUXHVZQI-UHFFFAOYSA-N
SMILES
O=C(CC1=CC=CC=C1)C1=CC=CC=C1
Form
Crystals or powder or crystalline powder or granules
Identification (FTIR)
Conforms
Appearance (Color)
White to pale cream
Melting Point (clear melt)
49.0-58.0?C
Assay (GC)
≥96.0%

Description

Deoxybenzoin, is used as a benzoin derivative and as a photoinitiator in vinyl polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Deoxybenzoin, is used as a benzoin derivative and as a photoinitiator in vinyl polymerization.

Solubility
Soluble in water (partly), methanol, alcohols, and ketones.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text