Thermo Scientific Chemicals

1-Ethyl-2-methylquinolinium iodide, 97%, Thermo Scientific Chemicals

Catalog number: A10584.06
5 g, Each
Thermo Scientific Chemicals

1-Ethyl-2-methylquinolinium iodide, 97%, Thermo Scientific Chemicals

Catalog number: A10584.06
5 g, Each
Quantity
Catalog number: A10584.06
also known as A10584-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
606-55-3
IUPAC Name
1-ethyl-2-methylquinolin-1-ium iodide
Molecular Formula
C12H14IN
InChI Key
OEVSHJVOKFWBJY-UHFFFAOYSA-M
SMILES
[I-].CC[N+]1=C2C=CC=CC2=CC=C1C
Form
Powder
Appearance (Color)
Yellow to brown
Assay (Titration ex Iodide)
≥96.0 to ≤104.0%

Description

1-Ethyl-2-methylquinolinium iodide is used as a precursor in synthesis of carbocyanine dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethyl-2-methylquinolinium iodide is used as a precursor in synthesis of carbocyanine dyes.

Solubility
Soluble in Water.

Notes
Precursor in synthesis of carbocyanine dyes.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text