Thermo Scientific Chemicals

DL-Leucine, 99%

Catalog number: A10590.18
50 g, Each
Thermo Scientific Chemicals

DL-Leucine, 99%

Catalog number: A10590.18
50 g, Each
Quantity
Catalog number: A10590.18
also known as A10590-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
328-39-2
IUPAC Name
2-amino-4-methylpentanoic acid
Molecular Formula
C6H13NO2
InChI Key
ROHFNLRQFUQHCH-UHFFFAOYNA-N
SMILES
CC(C)CC(N)C(O)=O
Appearance (Color)
White
Optical Rotation
0.0 ± 0.2? (c=1 in water)
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Form
Crystals or powder or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text