Thermo Scientific Chemicals

5-Nitro-2-furoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A10597.22
100 g, Each
Thermo Scientific Chemicals

5-Nitro-2-furoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A10597.22
100 g, Each
Quantity
Catalog number: A10597.22
also known as A10597-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
645-12-5
IUPAC Name
5-nitrofuran-2-carboxylic acid
Molecular Formula
C5H3NO5
InChI Key
IODMEDPPCXSFLD-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(O1)[N+]([O-])=O
Melting Point (clear melt)
183.0-189.0?C
Appearance (Color)
White to cream to yellow to brown
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Form
Crystals or powder or crystalline powder

Description

5-Nitro-2-furoic acid was used in the preparation of [1,2,4]triazolo[3,4-b][1,3,4]thiadiazines and [1,2,4]triazolo[3,4-b][1,3,4] thiadiazoles. The FT-IR and FT-Raman spectra of 5-Nitro-2-furoic acid has also been studied.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nitro-2-furoic acid was used in the preparation of [1,2,4]triazolo[3,4-b][1,3,4]thiadiazines and [1,2,4]triazolo[3,4-b][1,3,4] thiadiazoles. The FT-IR and FT-Raman spectra of 5-Nitro-2-furoic acid has also been studied.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text