Thermo Scientific Chemicals

1-Bromododecane, 98%, Thermo Scientific Chemicals

Catalog number: A10600.0I
5000 g, Each
Thermo Scientific Chemicals

1-Bromododecane, 98%, Thermo Scientific Chemicals

Catalog number: A10600.0I
5000 g, Each
Quantity
Catalog number: A10600.0I
also known as A10600-0I
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
143-15-7
IUPAC Name
1-bromododecane
Molecular Formula
C12H25Br
InChI Key
PBLNBZIONSLZBU-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCBr
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.5%
Refractive Index
1.4560-1.4600 @ 20?C

Description

1-Bromododecane is used as a solvent. It is used as an intermediate to manufacture surfactant, pharmaceuticals and other organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromododecane is used as a solvent. It is used as an intermediate to manufacture surfactant, pharmaceuticals and other organic compounds.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text