5-Bromo-2-chlorotoluene, 98%, Thermo Scientific Chemicals
5-Bromo-2-chlorotoluene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Bromo-2-chlorotoluene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number A10602.22
also known as A10602-22
Price (USD)/ Each
598.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
598.00
Add to cart
5-Bromo-2-chlorotoluene, 98%, Thermo Scientific Chemicals
Catalog numberA10602.22
Price (USD)/ Each
598.00
-
Add to cart
Chemical Identifiers
CAS54932-72-8
IUPAC Name4-bromo-1-chloro-2-methylbenzene
Molecular FormulaC7H6BrCl
InChI KeyOZFQMHJKAODEON-UHFFFAOYSA-N
SMILESCC1=C(Cl)C=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.5725-1.5765 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only