DL-Threonine, 99%
Thermo Scientific Chemicals

DL-Threonine, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A10606.14
also known as A10606-14
Price (USD)/ Each
43.30
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
43.30
Add to cart
DL-Threonine, 99%
Catalog numberA10606.14
Price (USD)/ Each
43.30
-
Add to cart
Chemical Identifiers
CAS80-68-2
IUPAC Name(2R,3R)-2-amino-3-hydroxybutanoic acid
Molecular FormulaC4H9NO3
InChI KeyAYFVYJQAPQTCCC-PWNYCUMCSA-N
SMILESC[C@@H](O)[C@@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale cream
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
DL-Threonine is used as a polar essential amino acid. It serves as a precursor of glycine as well as used in the biosynthesis of proteins. Further, it is used for biochemical research. In addition to this, it is used as a nutrition enhancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Threonine is used as a polar essential amino acid. It serves as a precursor of glycine as well as used in the biosynthesis of proteins. Further, it is used for biochemical research. In addition to this, it is used as a nutrition enhancer.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Aizawa, S. I.; Kidani, T.; Takada, S.; Ofusa, Y. Simple Resolution of Enantiomeric NMR Signals of alpha-Amino Acids by Using Samarium(III) Nitrate With L-Tartarate. Chirality 2015, 27 (5), 353-357.
  2. Han, S. W.; Shin, J. S. Preparation of d-threonine by biocatalytic kinetic resolution. J. Mol. Catal., B Enzym. 2015, 122, 227-232.