5-Nitroindole, 99%, Thermo Scientific Chemicals
5-Nitroindole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Nitroindole, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number A10608.06
also known as A10608-06
Price (USD)/ Each
102.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
102.00
Add to cart
5-Nitroindole, 99%, Thermo Scientific Chemicals
Catalog numberA10608.06
Price (USD)/ Each
102.00
-
Add to cart
Chemical Identifiers
CAS6146-52-7
IUPAC Name5-nitro-1H-indole
Molecular FormulaC8H6N2O2
InChI KeyOZFPSOBLQZPIAV-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C2NC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)Yellow
FormCrystals or powder or crystalline powder
Melting Point (clear melt)137.0-144.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only