5-Methyl-1H-benzotriazole, 98+%, Thermo Scientific Chemicals
5-Methyl-1H-benzotriazole, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Methyl-1H-benzotriazole, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number A10615.36
also known as A10615-36
Price (USD)/ Each
895.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
895.00
Add to cart
5-Methyl-1H-benzotriazole, 98+%, Thermo Scientific Chemicals
Catalog numberA10615.36
Price (USD)/ Each
895.00
-
Add to cart
Chemical Identifiers
CAS136-85-6
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (GC)≥98.0%
Appearance (Color)White to pale cream to pale brown
5-Methyl-1H-benzotriazole was used in determination of benzothiazoles and benzotriazoles in waste water samples by GC-MS. It is also used as a potential nitrification inhibitor of urea fertilizer in agricultural soils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methyl-1H-benzotriazole was used in determination of benzothiazoles and benzotriazoles in waste water samples by GC-MS. It is also used as a potential nitrification inhibitor of urea fertilizer in agricultural soils.

Solubility
Soluble in chloroform, methanol, water (6.0 g/L at 25°C), ethanol, and toluene (slightly).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Devon A.Cancilla; J.Christopher Baird, Steven W.Geis; Steven R.Corsi. Studies of the environmental fate and effect of aircraft deicing fluids: Detection of 5-methyl-1H-benzotriazole in the fathead minnow (Pimephales promelas). Environmental Toxicology and Chemistry. 2003, 22, (1),134-140
  2. Alexander Muller; Stefan C.Weiss; Judith Beißwenger; H.Georg Leukhardt; Wolfgang Schulz ; Wolfram Seitz; Wolfgang K.L.Ruck, Walter H.Weber. Identification of ozonation by-products of 4- and 5-methyl-1H-benzotriazole during the treatment of surface water to drinking water . Water Research. 2012, 46, (3),679-690