Thermo Scientific Chemicals

1-Nitroso-2-naphthol, 98%, Thermo Scientific Chemicals

Catalog number: A10622.09
10 g, Each
Thermo Scientific Chemicals

1-Nitroso-2-naphthol, 98%, Thermo Scientific Chemicals

Catalog number: A10622.09
10 g, Each
Quantity
Catalog number: A10622.09
also known as A10622-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
131-91-9
IUPAC Name
1-nitrosonaphthalen-2-ol
Molecular Formula
C10H7NO2
InChI Key
YXAOOTNFFAQIPZ-UHFFFAOYSA-N
SMILES
OC1=CC=C2C=CC=CC2=C1N=O
Appearance (Color)
Brown to dark brown
Assay (HPLC)
≥97.5%
Melting Point (clear melt)
104-110?C
Form
Powder
Identification (FTIR)
Conforms

Description

1-Nitroso-2-naphthol as a complexing agent for the on-line preconcentration of cobalt with FI-FAAS. 1-Nitroso-2-naphthol is the chelating ion-exchanger in the synthesis of alumina adsorbents that are of acidic, basic and neutral nature. It is used for the removal and preconcentration of Pb(II), Cu(II), Cr(III) from waste, as well as drinking water. It is also used in the preconcentration of cobalt using C(18) microcolumn with flow injection that is coupled to a flame atomic absorption spectrometry (FI-FAAS) system. 1-Nitroso-2-naphthol is used in the preparation of 1-nitroso-2-naphthol-sodium nitrite reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Nitroso-2-naphthol as a complexing agent for the on-line preconcentration of cobalt with FI-FAAS. 1-Nitroso-2-naphthol is the chelating ion-exchanger in the synthesis of alumina adsorbents that are of acidic, basic and neutral nature. It is used for the removal and preconcentration of Pb(II), Cu(II), Cr(III) from waste, as well as drinking water. It is also used in the preconcentration of cobalt using C(18) microcolumn with flow injection that is coupled to a flame atomic absorption spectrometry (FI-FAAS) system. 1-Nitroso-2-naphthol is used in the preparation of 1-nitroso-2-naphthol-sodium nitrite reagent.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text